Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Flu/RSV/SARS-CoV-2 Positive Control
SDP

Catalog No. 22158382 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-382 6 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-382 Supplier ZeptoMetrix Supplier No. NATFRC6C
Add to Cart
Add to Cart

Qualitative molecular control

NATtrol products are ready to use, inactivated full process controls designed to evaluate performance of molecular tests. They can be used for verification of assays, training of laboratory personnel and to monitor assay-kit lot performance. NATtrol products contain intact organisms and should be run in a manner similar to clinical specimens.

Specifications

Content And Storage 2°C to 8°C
Description Designed to evaluate the performance of nucleic acid tests for determination of the presence of respiratory virus nucleic acids.
Quantity 6 x 0.5 mL
Regulatory Status RUO
Strain Designations Flu A (A/Brisbane/10/07), Flu B (B/Florida/10/06), RSV B (CH93(18)-18) and SARS-COV-2 (USA-WA1/20220)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.