Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Fluo-8 AM, green fluorescent calcium binding dye, 1 mg, solid, soluble in DMSO, excitation 490 nm, emission 520 nm, medium calcium affinity (Kd 390 nM)

Catalog No. NC1106298
Encompass_Preferred

Fluo-8 AM is a cell-permeable, medium-affinity green fluorescent calcium-binding dye specifically designed for detecting intracellular calcium levels with high sensitivity and robustness. This dye displays a significant enhancement in fluorescence intensity when bound to Ca2+ (Kd = 390 nM) and offers a fluorescence signal that is 1.5 times brighter than Fluo-4 in cellular studies. With optimal excitation and emission wavelengths at 490 nm and 520 nm respectively, Fluo-8 AM is ideal for live imaging of cytosolic calcium in various functional assays in scientific research.

  • Cell-permeable dye
  • Medium calcium affinity (Kd = 390 nM)
  • Fluorescent color: Green
  • Excitation maximum: 490 nm
  • Emission maximum: 520 nm
  • Soluble in DMSO
  • Physical form: Solid
  • Storage temp: -20°C
  • Stable up to 12 months
  • 1.5x brighter than Fluo-4
  • Cited in over 50 publications

Catalog No. NC1106298 Supplier Abcam Supplier No. AB1427731MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluo-8 AM is a cell-permeable, medium-affinity green fluorescent calcium-binding dye specifically designed for detecting intracellular calcium levels with high sensitivity and robustness. This dye displays a significant enhancement in fluorescence intensity when bound to Ca2+ (Kd = 390 nM) and offers a fluorescence signal that is 1.5 times brighter than Fluo-4 in cellular studies. With optimal excitation and emission wavelengths at 490 nm and 520 nm respectively, Fluo-8 AM is ideal for live imaging of cytosolic calcium in various functional assays in scientific research.

  • Cell-permeable dye
  • Medium calcium affinity (Kd = 390 nM)
  • Fluorescent color: Green
  • Excitation maximum: 490 nm
  • Emission maximum: 520 nm
  • Soluble in DMSO
  • Physical form: Solid
  • Storage temp: -20°C
  • Stable up to 12 months
  • 1.5x brighter than Fluo-4
  • Cited in over 50 publications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.