Learn More
AdipoGen Fluazifop
SDP
Supplier:  AdipoGen CDXF0209M010
Encompass_Preferred
Chemical. CAS 69335-91-7. Formula C15H12F3NO4. MW 327.26. Synthetic. Phenoxy herbicide. Compound can be used as analytical reference material. Grass-selective herbicide that acts by inhibiting fatty acid synthesis de novo in sensitive species. Mode of action is through acetyl-CoA carboxylase ACC inhibition.
Catalog No. 50-256-0867