Learn More
AdipoGen Fluazifop Solution (2 ml)
SDP
Supplier:  AdipoGen CDXF0341L002
Encompass_Preferred
Chemical. CAS: 69335-91-7. Formula: C15H12F3NO4. MW: 327.26. Phenoxy herbicide. Compound can be used as analytical reference material. Grass-selective herbicide that acts by inhibiting fatty acid synthesis de novo in sensitive species. Mode of action is through acetyl-CoA carboxylase (ACC) inhibition.
Catalog No. 50-205-4452