Learn More
Medchemexpress LLC Fluazinam | 79622-59-6 | MFCD00214168 | 99.3% | 465.09 g/mol | C13H4Cl2F6N4O4 | 10 MG
SDP
Catalog No. 5000297132
Encompass_Preferred
Fluazinam (Standard) is an analytical reference standard of the fungicide fluazinam supplied as a solid for laboratory research and analytical purposes. With molecular formula C13H4Cl2F6N4O4 and a molecular weight of 465.09 g/mol, this 10 mg package provides a high-purity material for assay development, calibration, and method validation.
  • High purity: 99.3% (rounded assay value).
  • Suitable as an analytical standard for method development and calibration.
  • Supplied as a solid in a 10 mg quantity for small-scale analytical use.
  • Safety data sheet available for handling and storage guidance.
Catalog No. 50-002-97132 Supplier Medchemexpress LLC Supplier No. HYB1839R10MG
Fluazinam (Standard) is an analytical reference standard of the fungicide fluazinam supplied as a solid for laboratory research and analytical purposes. With molecular formula C13H4Cl2F6N4O4 and a molecular weight of 465.09 g/mol, this 10 mg package provides a high-purity material for assay development, calibration, and method validation.
  • High purity: 99.3% (rounded assay value).
  • Suitable as an analytical standard for method development and calibration.
  • Supplied as a solid in a 10 mg quantity for small-scale analytical use.
  • Safety data sheet available for handling and storage guidance.