Learn More
Medchemexpress LLC Fluazinam (Standard) | 79622-59-6 | 99.3% | 100 MG
SDP
Catalog No. 5000412369
Encompass_Preferred
Fluazinam is a broad-spectrum pyridinamine fungal inhibitor and an orally active dinitroaniline fungicide. It is intended for research and analytical applications, and is commonly used as an analytical standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Induces phosphorylation of JNK
  • Activates p38 pathway
  • Decreases Bcl-2
  • Activates caspase-3
  • Decreases complex I activity
  • Increases autophagy and apoptosis
  • Exhibits strong antifungal activity against F. fujikuroi and B. maydis
  • Intended for research and analytical applications
  • Functions as an analytical standard for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-12369 Supplier Medchemexpress LLC Supplier No. HYB1839R100MG
Fluazinam is a broad-spectrum pyridinamine fungal inhibitor and an orally active dinitroaniline fungicide. It is intended for research and analytical applications, and is commonly used as an analytical standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Induces phosphorylation of JNK
  • Activates p38 pathway
  • Decreases Bcl-2
  • Activates caspase-3
  • Decreases complex I activity
  • Increases autophagy and apoptosis
  • Exhibits strong antifungal activity against F. fujikuroi and B. maydis
  • Intended for research and analytical applications
  • Functions as an analytical standard for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS