Learn More
Medchemexpress LLC Flubendazole-d3 | 1173021-08-3 | 96.00% | 1 MG
SDP
Catalog No. 5000406446
Encompass_Preferred
Flubendazole-d3 is a deuterium-labeled version of Flubendazole, a safe and effective anthelmintic agent used in humans, rodents, and ruminants. It demonstrates anticancer activities by inhibiting microtubule function, inducing p53-mediated apoptosis, and arresting the G2/M cell cycle. Deuterium substitution in drug molecules can impact their pharmacokinetic and metabolic profiles.
  • Deuterium labeled version of Flubendazole
  • Safe and effective anthelmintic agent
  • Exhibits anticancer activities via microtubule inhibition
  • Induces p53-mediated apoptosis
  • Arrests G2/M cell cycle
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis (e.g., NMR, GC-MS, LC-MS)
Catalog No. 50-004-06446 Supplier Medchemexpress LLC Supplier No. HYB0294S1MG
Flubendazole-d3 is a deuterium-labeled version of Flubendazole, a safe and effective anthelmintic agent used in humans, rodents, and ruminants. It demonstrates anticancer activities by inhibiting microtubule function, inducing p53-mediated apoptosis, and arresting the G2/M cell cycle. Deuterium substitution in drug molecules can impact their pharmacokinetic and metabolic profiles.
  • Deuterium labeled version of Flubendazole
  • Safe and effective anthelmintic agent
  • Exhibits anticancer activities via microtubule inhibition
  • Induces p53-mediated apoptosis
  • Arrests G2/M cell cycle
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis (e.g., NMR, GC-MS, LC-MS)