Learn More
Biotium FLUBIDA-2
Catalog No. 501964709
Encompass_Preferred
Flubida-2 has been used to detect pH at a specific site in a cell such as cell organelles by directing the probe to where avidin fusion proteins are located. The probe is a conjugate of biotin and fluorescein diacetate, which is nonfluorescent until the probe has entered the cells and hydrolyzed by endoesterases. Flubida-2 is membrane-permeable and thus can be delivered into cells via simple incubation with the probe in a buffer. Properties: Off-white solid soluble in DMSO; Store at 4°C; C43H48N4O9S; MW: 812.93
Catalog No. 50-196-4709 Supplier Biotium Supplier No. 51022
Flubida-2 has been used to detect pH at a specific site in a cell such as cell organelles by directing the probe to where avidin fusion proteins are located. The probe is a conjugate of biotin and fluorescein diacetate, which is nonfluorescent until the probe has entered the cells and hydrolyzed by endoesterases. Flubida-2 is membrane-permeable and thus can be delivered into cells via simple incubation with the probe in a buffer. Properties: Off-white solid soluble in DMSO; Store at 4°C; C43H48N4O9S; MW: 812.93