Learn More
Cayman Chemical FlucloxacIlInsodIum salt 100mg
SDP
Catalog No. 502913678
Encompass_Preferred
A β-lactam antibiotic; active against clinical isolates of S. aureus (MICs = 0.12-0.25 mg/L); decreases vegetation titers in a rat model of S. aureus-induced aortic valve endocarditis at 42.3 mg/kg every six hours
Catalog No. 50-291-3678 Supplier Cayman Chemical Supplier No. 35241100 MG
A β-lactam antibiotic; active against clinical isolates of S. aureus (MICs = 0.12-0.25 mg/L); decreases vegetation titers in a rat model of S. aureus-induced aortic valve endocarditis at 42.3 mg/kg every six hours