Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flucloxacillin sodium | 1847-24-1 | 98.0% | C19H16ClFN3NaO5S | 1 G
SDP

Catalog No. 5000432384
Encompass_Preferred

Flucloxacillin sodium is an active antibiotic that effectively targets both gram-positive and gram-negative bacteria. With a purity of 98.02%, it is commonly used in research and has shown efficacy in reducing germs in biofilm and bone tissue in periprosthetic infections. This compound is also being investigated in various clinical trials for conditions such as cellulitis, wound infection, and osteomyelitis.

  • Active against gram-positive and gram-negative bacteria
  • Effective in periprosthetic infection models
  • Reduces germs in biofilm and bone tissue
  • Purity of 98.02%
  • Soluble in water and DMSO
  • Involved in clinical trials for various bacterial infections

Catalog No. 50-004-32384 Supplier Medchemexpress LLC Supplier No. HYA0246A1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flucloxacillin sodium is an active antibiotic that effectively targets both gram-positive and gram-negative bacteria. With a purity of 98.02%, it is commonly used in research and has shown efficacy in reducing germs in biofilm and bone tissue in periprosthetic infections. This compound is also being investigated in various clinical trials for conditions such as cellulitis, wound infection, and osteomyelitis.

  • Active against gram-positive and gram-negative bacteria
  • Effective in periprosthetic infection models
  • Reduces germs in biofilm and bone tissue
  • Purity of 98.02%
  • Soluble in water and DMSO
  • Involved in clinical trials for various bacterial infections

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.