Learn More
Medchemexpress LLC Flucloxacillin sodium | 1847-24-1 | 98.0% | C19H16ClFN3NaO5S | 500 MG
SDP
Catalog No. 5000432266
Encompass_Preferred
Flucloxacillin sodium is an active antibiotic effective against both gram-positive and gram-negative bacteria, intended for research use only.
  • Active antibiotic
  • Effective against gram-positive and gram-negative bacteria
  • Demonstrated effectiveness in research against periprosthetic infection models
Catalog No. 50-004-32266 Supplier Medchemexpress LLC Supplier No. HYA0246A500MG
Flucloxacillin sodium is an active antibiotic effective against both gram-positive and gram-negative bacteria, intended for research use only.
  • Active antibiotic
  • Effective against gram-positive and gram-negative bacteria
  • Demonstrated effectiveness in research against periprosthetic infection models