Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flucloxacillin Sodium, G-Biosciences
SCI_ED

Catalog No. S139779
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139779 10 mg
S139084 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139779 Supplier G-Biosciences Supplier No. RC1023
Add to Cart
Add to Cart

Flucloxacillin sodium is a narrow-spectrum penicillin antibiotic similar to cloxacillin.

Resistant to Beta-lactamases, it targets gram-positive organisms such as Staphylococcus aureus.

Chemical Identifiers

Molecular Weight (g/mol) 475.85 g/mol

Specifications

Boiling Point 677°C
Quantity 10 mg
Percent Purity 98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.