Learn More
Medchemexpress LLC Flucofuron | 370-50-3 | 417.13 | 250 MG
SDP
Catalog No. 5000365768
Encompass_Preferred
Flucofuron is a highly effective compound against *N. fowleri* trophozoites and the resistant cyst stage. It can induce cell apoptosis and is suitable for research in infection and inflammation, including Primary Amoebic Meningoencephalitis (PAM).
  • High efficacy against *N. fowleri* trophozoites (IC50 values of 2.58 μM for ATCC 30808 and 2.47 μM for ATCC 30215).
  • Active against resistant cyst stage (IC50 = 0.88 μM).
  • Induces cell apoptosis.
  • Useful for infection and inflammation research.
Catalog No. 50-003-65768 Supplier Medchemexpress LLC Supplier No. 250MGHYI0673
Flucofuron is a highly effective compound against *N. fowleri* trophozoites and the resistant cyst stage. It can induce cell apoptosis and is suitable for research in infection and inflammation, including Primary Amoebic Meningoencephalitis (PAM).
  • High efficacy against *N. fowleri* trophozoites (IC50 values of 2.58 μM for ATCC 30808 and 2.47 μM for ATCC 30215).
  • Active against resistant cyst stage (IC50 = 0.88 μM).
  • Induces cell apoptosis.
  • Useful for infection and inflammation research.