Learn More
Selleck Chemical LLC Fluconazole S1331-1g
SDP
Catalog No. 503110359
Encompass_Preferred
Fluconazole is a fungal lanosterol 14 alpha-demethylase inhibitor which thereby prevents the formation of ergosterol used in the treatment and prevention of superficial and systemic fungal infections
Catalog No. 50-311-0359 Supplier Selleck Chemical LLC Supplier No. S13311G
Fluconazole is a fungal lanosterol 14 alpha-demethylase inhibitor which thereby prevents the formation of ergosterol used in the treatment and prevention of superficial and systemic fungal infections