Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Fluconazole, 98%

Catalog No. AC455480050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455480050 5 g
AC455480010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455480050 Supplier Thermo Scientific Chemicals Supplier No. 455480050
  • $436.00
    $351.65 / Each of 1

    Save $84.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Fluconazole, 98%, CAS # 86386-73-4, is a synthetic triazole with antifungal activity against some species of Candida. It also shows inhibitory activity against cytochrome P-450 enzymes and it is a demethylase. | CAS: 86386-73-4 | C13H12F2N6O | 306.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Fluconazole, 98% is a triazole compound showing antifungal activity mainly against Candida, Coccidioides, and Cryptococcus species

• It has been shown to inhibit fungal cytochrome P-450 sterol C-14 α-demethylation, resulting in the accumulation of fungal 14 α-methyl sterols, the loss of normal fungal sterols, and associated fungistatic activity

• It also inhibits cytochrome P450 family 2 subfamily c member 9 (CYP2C9)

• Mammalian cell demethylation is much less susceptible to fluconazole inhibition

Applications

• Fluconazole has fungistatic activity against many fungal strains including Candida albicans, Candida glabrata, Candida parapsilosis, Candida tropicalis, and Cryptococcus neoformans.

• Fluconazole interferes with fungal ergosterol synthesis and downregulates the metallothionein gene

• In biological experiments, this compound has been used to assess minimum inhibitory concentration in (MIC)-fungal assays

• It has also been used as a positive control in antifungal assay of plants

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86386-73-4
Molecular Formula C13H12F2N6O
Molecular Weight (g/mol) 306.27
InChI Key RFHAOTPXVQNOHP-UHFFFAOYSA-N
Synonym fluconazole, diflucan, triflucan, elazor, biozolene, biocanol, fungata, fluconazol, fluconazolum, flucazol
PubChem CID 3365
ChEBI CHEBI:46081
IUPAC Name 2-(2,4-difluorophenyl)-1,3-bis(1,2,4-triazol-1-yl)propan-2-ol
SMILES C1=CC(=C(C=C1F)F)C(CN2C=NC=N2)(CN3C=NC=N3)O

Specifications

Melting Point 137.0°C to 141.0°C
Loss on Drying 1% max. (105°C, 3 h)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: <1mg/mL at 25 deg c. Other solubilities: solubility in ethanol: 61mg/mL at 25 deg c, solubility in dmso: 61mg/mL at 25 deg c
Formula Weight 306.27
Percent Purity 98%
Chemical Name or Material Fluconazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May damage the unborn child.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.