Learn More
Medchemexpress LLC Fluconazole-d4 | 1124197-58-5 | 99.03% | C13H8D4F2N6O | 1 MG
SDP
Catalog No. 5000433354
Encompass_Preferred
Fluconazole-d4 is a deuterium-labeled Fluconazole, a triazole antifungal agent effective against various fungi, especially *Candida albicans*. It inhibits *C. albicans* and *Candida kefyr* with IC99s ranging from 0.20 μg/mL to 0.39 μg/mL. Deuterium labeling allows its use as a tracer in drug development, potentially influencing pharmacokinetic and metabolic profiles.
  • Used as a tracer in drug development
  • Functions as an internal standard for quantitative analysis
  • Applicable in techniques like NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-33354 Supplier Medchemexpress LLC Supplier No. HYB0101S1MG
Fluconazole-d4 is a deuterium-labeled Fluconazole, a triazole antifungal agent effective against various fungi, especially *Candida albicans*. It inhibits *C. albicans* and *Candida kefyr* with IC99s ranging from 0.20 μg/mL to 0.39 μg/mL. Deuterium labeling allows its use as a tracer in drug development, potentially influencing pharmacokinetic and metabolic profiles.
  • Used as a tracer in drug development
  • Functions as an internal standard for quantitative analysis
  • Applicable in techniques like NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs