Learn More
Cayman Chemical Fluconazole-d4 1mg
SDP
Catalog No. 502965809
Encompass_Preferred
An internal standard for the quantification of fluconazole by GC- or LC-MS
Catalog No. 50-296-5809 Supplier Cayman Chemical Supplier No. 182521 MG
Add to Cart
Add to Cart
An internal standard for the quantification of fluconazole by GC- or LC-MS