Learn More
Cayman Chemical Fluconazole-d4 500ug
SDP
Catalog No. 502965725
Encompass_Preferred
An internal standard for the quantification of fluconazole by GC- or LC-MS
Catalog No. 50-296-5725 Supplier Cayman Chemical Supplier No. 18252500 UG
Add to Cart
Add to Cart
An internal standard for the quantification of fluconazole by GC- or LC-MS