Learn More
Apexbio Technology LLC Fluconazole hydrate 155347-36-7 5g
SDP
Catalog No. 5000092165
Encompass_Preferred
Fluconazole hydrate (CAS 155347-36-7) is a small-molecule inhibitor targeting the fungal enzyme lanosterol 14 -demethylase It is designed to inhibit ergosterol biosynthesis thereby disrupting fungal cell membrane integrity and arresting fungal growth Fluconazole hydrate exerts its biological activity primarily through inhibition of lanosterol 14 -demethylase In in vitro studies fluconazole hydrate demonstrates inhibitory activity with documented IC50 values typically ranging from 0 1 to 10 g/mL depending on fungal strain and experimental conditions Based on these pharmacological properties fluconazole hydrate holds research potential in antifungal pharmacology assays fungal infection model studies and evaluation of combinational antifungal therapies
Catalog No. 50-000-92165 Supplier Apexbio Technology LLC Supplier No. B12265000
Fluconazole hydrate (CAS 155347-36-7) is a small-molecule inhibitor targeting the fungal enzyme lanosterol 14 -demethylase It is designed to inhibit ergosterol biosynthesis thereby disrupting fungal cell membrane integrity and arresting fungal growth Fluconazole hydrate exerts its biological activity primarily through inhibition of lanosterol 14 -demethylase In in vitro studies fluconazole hydrate demonstrates inhibitory activity with documented IC50 values typically ranging from 0 1 to 10 g/mL depending on fungal strain and experimental conditions Based on these pharmacological properties fluconazole hydrate holds research potential in antifungal pharmacology assays fungal infection model studies and evaluation of combinational antifungal therapies