Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluconazole Impurity Standard, MilliporeSigma™ Supelco™

Catalog No. 111001858 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-1858 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-1858 Supplier MilliporeSigma Supelco Supplier No. BP1175
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
TRUSTED_SUSTAINABILITY

Specifications

CAS 86386-73-4
Linear Formula C13H12F2N6 O
MDL Number MFCD00274549
Synonym Fluconazole; 2-(2,4-Difluorophenyl)-1,3-bis(1 H-1,2,4-triazol-1-yl)propan-2-ol
Type Pharmacopeia and Metrological Institutes Standards
Quantity 25 mg
Formula Weight 306.27
Grade Pharmaceutical Primary Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.