Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluconazole, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602741 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-741 25 g Amber Glass Bottle
18-602-742 100 g Amber Glass Bottle
18-602-746 500 g Amber Glass Bottle
18-602-743 1 kg Amber Glass Bottle
18-602-744 5 kg Amber Glass Bottle
18-602-745 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-602-741 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F135525GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

F1355, 86386-73-4, C13H12F2N6O

Spectrum™ Chemical Fluconazole, USP is used to treat and prevent superficial and systematic fungal infections. Known as a triazole antifungal drug, it has a white crystalline appearance and is slightly soluble in water and alcohol. Most commonly found under the trade names Diflucan and Trican and is listed on the World Health Organizations list of essential medicines. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 86386-73-4
Molecular Formula C13H12F2N6O
Molecular Weight (g/mol) 306.28
InChI Key RFHAOTPXVQNOHP-UHFFFAOYSA-N
IUPAC Name 2-(2,4-difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol
SMILES OC(CN1C=NC=N1)(CN1C=NC=N1)C1=CC=C(F)C=C1F

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.