Learn More
Apexbio Technology LLC Fludarabine Phosphate (Fludara)(Synonyms: Fludara, Fludarabine phosphate, Fludarabine monophosphate, 2-Fluoro-ara-AMP), 200mg, CAS: 75607-67-9.
SDP
Catalog No. 5000060935
Encompass_Preferred
Fludarabine phosphate (CAS 75607-67-9) marketed as Fludara is a fluorinated nucleotide analog derived from vidarabine (ara-A) exhibiting antineoplastic activities by interfering with DNA synthesis Following rapid extracellular dephosphorylation to 2-fluoro-ara-A this metabolite is intracellularly phosphorylated by deoxycytidine kinase generating the active triphosphate metabolite 2-fluoro-ara-ATP The active metabolite inhibits critical enzymes including DNA polymerase alpha ribonucleotide reductase and DNA primase thereby disrupting DNA replication and ultimately impeding tumor cell proliferation This compound is broadly utilized in biomedical research focused on anticancer mechanisms and nucleotide metabolism studies
Catalog No. 50-000-60935 Supplier Apexbio Technology LLC Supplier No. A8317200
Fludarabine phosphate (CAS 75607-67-9) marketed as Fludara is a fluorinated nucleotide analog derived from vidarabine (ara-A) exhibiting antineoplastic activities by interfering with DNA synthesis Following rapid extracellular dephosphorylation to 2-fluoro-ara-A this metabolite is intracellularly phosphorylated by deoxycytidine kinase generating the active triphosphate metabolite 2-fluoro-ara-ATP The active metabolite inhibits critical enzymes including DNA polymerase alpha ribonucleotide reductase and DNA primase thereby disrupting DNA replication and ultimately impeding tumor cell proliferation This compound is broadly utilized in biomedical research focused on anticancer mechanisms and nucleotide metabolism studies