Learn More
Cayman Chemical FludarabIn Phosphate 10mg
SDP
Catalog No. 502955340
Encompass_Preferred
A prodrug form of fludarabine and 2-fluoro-ara-ATP; reduces viability of, and induces apoptosis in, N-Myc-overexpressing LASCPC-01 cells (IC50 = 17.67 µM); reduces N-Myc protein levels when used at 25 µM for at least 48 hours; reduces tumor volume in N-Myc-overexpressing 22Rv1-N-Myc and LASCPC-01, but not non-N-Myc-overexpressing 22Rv1, mouse xenograft models at 120 mg/kg; an inhibitor of SARS-CoV-2 RdRp (EC50 = 0.93 µM)
Catalog No. 50-295-5340 Supplier Cayman Chemical Supplier No. 1425110 MG
A prodrug form of fludarabine and 2-fluoro-ara-ATP; reduces viability of, and induces apoptosis in, N-Myc-overexpressing LASCPC-01 cells (IC50 = 17.67 µM); reduces N-Myc protein levels when used at 25 µM for at least 48 hours; reduces tumor volume in N-Myc-overexpressing 22Rv1-N-Myc and LASCPC-01, but not non-N-Myc-overexpressing 22Rv1, mouse xenograft models at 120 mg/kg; an inhibitor of SARS-CoV-2 RdRp (EC50 = 0.93 µM)