Learn More
Selleck Chemical LLC Fludarabine 10mM/1mL 21679-14-1 FaraA, Fludarabinum
SDP
Supplier:  Selleck Chemical LLC S149110MM/1ML
Encompass_Preferred
Fludarabine (FaraA, Fludarabinum) is a STAT1 activation inhibitor which causes a specific depletion of STAT1 protein (and mRNA) but not of other STATs. Also a DNA synthesis inhibitor in vascular smooth muscle cells. Fludarabine induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1287468