Learn More
Apexbio Technology LLC Fludarabine(Synonyms: Fludarabine Phosphate, Fludara, Fludarabinum, 2-Fluoro-ara-AMP, 9H-Purin-6-amine), 10mM (in 1mL DMSO), CAS: 21679-14-1.
SDP
Catalog No. 5000060964
Encompass_Preferred
Fludarabine (CAS 21679-14-1) is a purine analog prodrug that inhibits DNA synthesis Upon cellular uptake it undergoes phosphorylation to its active triphosphate form (F-ara-ATP) disrupting DNA replication through inhibition of critical enzymes such as DNA primase DNA ligase I ribonucleotide reductase and DNA polymerases and In human myeloma RPMI8226 cell assays fludarabine suppressed cell growth reduced Akt phosphorylation and lowered anti-apoptotic proteins XIAP and Survivin In vivo studies demonstrated marked tumor growth inhibition in RPMI8226 xenograft models Fludarabine serves as a tool in oncology research particularly leukemia and multiple myeloma-related studies
Catalog No. 50-000-60964 Supplier Apexbio Technology LLC Supplier No. A54245.1
Fludarabine (CAS 21679-14-1) is a purine analog prodrug that inhibits DNA synthesis Upon cellular uptake it undergoes phosphorylation to its active triphosphate form (F-ara-ATP) disrupting DNA replication through inhibition of critical enzymes such as DNA primase DNA ligase I ribonucleotide reductase and DNA polymerases and In human myeloma RPMI8226 cell assays fludarabine suppressed cell growth reduced Akt phosphorylation and lowered anti-apoptotic proteins XIAP and Survivin In vivo studies demonstrated marked tumor growth inhibition in RPMI8226 xenograft models Fludarabine serves as a tool in oncology research particularly leukemia and multiple myeloma-related studies