Learn More
Medchemexpress LLC HY-B0069 50mg , Fludarabine CAS:21679-14-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB006950MG
Encompass_Preferred
HY-B0069 50mg Fludarabine CAS: 21679-14-1 Fludarabine (NSC 118218) is a DNA synthesis inhibitor and a fluorinated purine analogue with antineoplastic activity in lymphoproliferative malignancies. Fludarabine inhibits the cytokine-induced activation of STAT1 and STAT1-dependent gene transcription in normal resting or activated lymphocytes[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1115