Learn More
Enzo Life Sciences Fludarabine (5mg). CAS: 21679-14-1
SDP
Supplier:  Enzo Life Sciences ALX480100M005
Encompass_Preferred
Prodrug converted to the free nucleoside 9-β-D-arabinosyl-2-fluoroadenine (F-ara-A). F-ara-A enters cells and accumulates as the 5'-triphosphate. Interferes with DNA synthesis and repair. Induces apoptosis of cancer cells. Inhibits RNA transcription. Alternative name: 2-Fluoroadenine-9-beta-D-arabinofuranoside. Purity: ≥98% (HPLC). Appearance: White to pale yellow crystalline powder. Solubility: Sparingly soluble in water or organic solvents. Long Term Storage: +4°C.
Catalog No. 50-200-9330