Learn More
Medchemexpress LLC HY-145443 1mg , Fludarabine-Cl CAS:2734853-80-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1454431MG
Encompass_Preferred
HY-145443 1mg Fludarabine-Cl CAS: 2734853-80-4 Fludarabine-Cl has inhibition effect on RNA adenosine deaminase 1(ADAR1), and can be used for preventing and/or treating cancer or tumor-related diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0867