Learn More
Medchemexpress LLC Fludarabine-Cl | 2734853-80-4 | 99.7% | C10H11ClFN5O3 | 10MG
SDP
Catalog No. 5000259850
Encompass_Preferred
Fludarabine-Cl is a small-molecule inhibitor of RNA adenosine deaminase 1 (ADAR1) used for research into RNA editing and cancer-related biology. Supplied as a high-purity solid, the compound has defined solubility and storage guidelines to support in vitro and in vivo experimentation.
  • Inhibits ADAR1 with an IC50 of 0.87 μM.
  • High purity (99.71%) suitable for research applications.
  • Available in multiple sizes including small milligram quantities.
  • Soluble in water (5 mg/mL) and PBS (2 mg/mL) with recommended ultrasonic assistance.
  • Powder stable at -20°C for extended storage; solutions stable at -80°C.
  • Supplied as a solid powder convenient for assay preparation.
Catalog No. 50-002-59850 Supplier Medchemexpress LLC Supplier No. HY14544310MG
Fludarabine-Cl is a small-molecule inhibitor of RNA adenosine deaminase 1 (ADAR1) used for research into RNA editing and cancer-related biology. Supplied as a high-purity solid, the compound has defined solubility and storage guidelines to support in vitro and in vivo experimentation.
  • Inhibits ADAR1 with an IC50 of 0.87 μM.
  • High purity (99.71%) suitable for research applications.
  • Available in multiple sizes including small milligram quantities.
  • Soluble in water (5 mg/mL) and PBS (2 mg/mL) with recommended ultrasonic assistance.
  • Powder stable at -20°C for extended storage; solutions stable at -80°C.
  • Supplied as a solid powder convenient for assay preparation.