Learn More
Abcam Fludarabine (FAMP), DNA synthesis inhibitor, 10MG
Catalog No. 502706951
Encompass_Preferred
MW 285.23 Da, Purity >98%. DNA synthesis inhibitor. Prodrug converted to active metabolite F-ara-A which inhibits ribonucleotide reductase and competitively inhibits DNA polymerase. Also prevents DNA ligase I from joining DNA. Induces apoptosis. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6951 Supplier Abcam Supplier No. AB14104810MG
MW 285.23 Da, Purity >98%. DNA synthesis inhibitor. Prodrug converted to active metabolite F-ara-A which inhibits ribonucleotide reductase and competitively inhibits DNA polymerase. Also prevents DNA ligase I from joining DNA. Induces apoptosis. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement