Learn More
Selleck Chemical LLC Fludarabine 50mg 21679-14-1 FaraA, Fludarabinum
SDP
Catalog No. 506777
Encompass_Preferred
Fludarabine (FaraA, Fludarabinum) is a STAT1 activation inhibitor which causes a specific depletion of STAT1 protein (and mRNA) but not of other STATs. Also a DNA synthesis inhibitor in vascular smooth muscle cells. Fludarabine induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-677-7 Supplier Selleck Chemical LLC Supplier No. S149150MG
Fludarabine (FaraA, Fludarabinum) is a STAT1 activation inhibitor which causes a specific depletion of STAT1 protein (and mRNA) but not of other STATs. Also a DNA synthesis inhibitor in vascular smooth muscle cells. Fludarabine induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.