Learn More
TARGETMOL CHEMICALS INC FLUDARABINE PHOSPHATE 50MG
SDP
Catalog No. 502576502
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fludarabine Phosphate (NSC 312887 Phosphate) is the phosphate salt of a fluorinated nucleotide antimetabolite analog of the antiviral agent vidarabine (ara-A) with antineoplastic activity. Fludarabine phosphate is rapidly dephosphorylated to 2-fluoro-ara-A and then phosphorylated intracellularly by deoxycytidine kinase to the active triphosphate 2-fluoro-ara-ATP. This metabolite may inhibit DNA polymerase alpha ribonucleotide reductase and DNA primase thereby interrupting DNA synthesis and inhibiting tumor cell growth. purity: 99%
Catalog No. 50-257-6502 Supplier TARGETMOL CHEMICALS INC Supplier No. T650150MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fludarabine Phosphate (NSC 312887 Phosphate) is the phosphate salt of a fluorinated nucleotide antimetabolite analog of the antiviral agent vidarabine (ara-A) with antineoplastic activity. Fludarabine phosphate is rapidly dephosphorylated to 2-fluoro-ara-A and then phosphorylated intracellularly by deoxycytidine kinase to the active triphosphate 2-fluoro-ara-ATP. This metabolite may inhibit DNA polymerase alpha ribonucleotide reductase and DNA primase thereby interrupting DNA synthesis and inhibiting tumor cell growth. purity: 99%