Learn More
Medchemexpress LLC Fludarabine (phosphate) | 75607-67-9 | 99.8% | C10H13FN5O7P | 1 ML
SDP
Catalog No. 5000432575
Encompass_Preferred
Fludarabine phosphate is an analogue of adenosine and deoxyadenosine, which competes with dATP for incorporation into DNA and inhibits DNA synthesis. This compound is for research use only and has not been fully validated for medical applications.
  • Significantly reduces cell viability in a dose-dependent manner
  • Leads to complete regressions of all tumors and cures of all mice in some in vivo studies
  • Used for research purposes
Catalog No. 50-004-32575 Supplier Medchemexpress LLC Supplier No. HYB002810MM/1ML
Fludarabine phosphate is an analogue of adenosine and deoxyadenosine, which competes with dATP for incorporation into DNA and inhibits DNA synthesis. This compound is for research use only and has not been fully validated for medical applications.
  • Significantly reduces cell viability in a dose-dependent manner
  • Leads to complete regressions of all tumors and cures of all mice in some in vivo studies
  • Used for research purposes