Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fludarabine phosphate | 75607-67-9 | 99.8% | C10H13FN5O7P | 5 MG
SDP

Catalog No. 5000432831
Encompass_Preferred

Fludarabine phosphate is an analogue of adenosine and deoxyadenosine that competes with dATP for incorporation into DNA, thereby inhibiting DNA synthesis. This compound is primarily used in laboratory research for studying its biochemical effects.

  • Inhibits DNA synthesis
  • White to off-white solid appearance
  • CAS number 75607-67-9
  • Purity of 99.77% by HPLC
  • Stable as powder for 3 years at -20°C or 2 years at 4°C
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C

Catalog No. 50-004-32831 Supplier Medchemexpress LLC Supplier No. HYB00285MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fludarabine phosphate is an analogue of adenosine and deoxyadenosine that competes with dATP for incorporation into DNA, thereby inhibiting DNA synthesis. This compound is primarily used in laboratory research for studying its biochemical effects.

  • Inhibits DNA synthesis
  • White to off-white solid appearance
  • CAS number 75607-67-9
  • Purity of 99.77% by HPLC
  • Stable as powder for 3 years at -20°C or 2 years at 4°C
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.