Learn More
Medchemexpress LLC Fludarabine (phosphate) | 75607-67-9 | 99.8% | C10H13FN5O7P | 50 MG
SDP
Catalog No. 5000432833
Encompass_Preferred
Fludarabine phosphate is an analogue of adenosine and deoxyadenosine. It competes with dATP for incorporation into DNA and inhibits DNA synthesis. This compound significantly reduces cell viability in a dose-dependent manner.
  • Competes with dATP for incorporation into DNA
  • Inhibits DNA synthesis
  • Significantly reduces cell viability in a dose-dependent manner
  • Causes complete regression of D54 tumors in mice
  • For research use only
Catalog No. 50-004-32833 Supplier Medchemexpress LLC Supplier No. HYB002850MG
Fludarabine phosphate is an analogue of adenosine and deoxyadenosine. It competes with dATP for incorporation into DNA and inhibits DNA synthesis. This compound significantly reduces cell viability in a dose-dependent manner.
  • Competes with dATP for incorporation into DNA
  • Inhibits DNA synthesis
  • Significantly reduces cell viability in a dose-dependent manner
  • Causes complete regression of D54 tumors in mice
  • For research use only