Learn More
Medchemexpress LLC Fludarabine triphosphate trisodium | 00-00-0 | 100.0% | C10H12FN5Na3O13P3 | 10MG
SDP
Catalog No. 5000259109
Encompass_Preferred
Fludarabine triphosphate trisodium is the active triphosphate metabolite of fludarabine that interferes with DNA synthesis. It inhibits DNA primase, ribonucleotide reductase, and DNA polymerase, leading to disruption of DNA replication and induction of apoptosis. Supplied as a high-purity trisodium salt in liquid form for research use.
  • Active triphosphate metabolite that inhibits DNA primase, ribonucleotide reductase, and DNA polymerase.
  • Disrupts DNA synthesis and promotes apoptosis in target cells.
  • High purity suitable for biochemical and cell-free assays.
  • Trisodium salt provided as a liquid, colorless to light yellow.
  • Storage conditions support stability when protected from light and stored under nitrogen.
Catalog No. 50-002-59109 Supplier Medchemexpress LLC Supplier No. HY136650A10MG
Fludarabine triphosphate trisodium is the active triphosphate metabolite of fludarabine that interferes with DNA synthesis. It inhibits DNA primase, ribonucleotide reductase, and DNA polymerase, leading to disruption of DNA replication and induction of apoptosis. Supplied as a high-purity trisodium salt in liquid form for research use.
  • Active triphosphate metabolite that inhibits DNA primase, ribonucleotide reductase, and DNA polymerase.
  • Disrupts DNA synthesis and promotes apoptosis in target cells.
  • High purity suitable for biochemical and cell-free assays.
  • Trisodium salt provided as a liquid, colorless to light yellow.
  • Storage conditions support stability when protected from light and stored under nitrogen.