Learn More
Medchemexpress LLC Fludarabine triphosphate trisodium | 100.0% | 591.12 | C10H12FN5Na3O13P3 | 5 MG
SDP
Catalog No. 5000221843
Encompass_Preferred
Fludarabine triphosphate trisodium is the active triphosphate metabolite of fludarabine that inhibits DNA primase and interferes with DNA synthesis. It blocks primer RNA formation, inhibits ribonucleotide reductase and DNA polymerase, and promotes apoptosis in susceptible cells. Supplied as a colorless to light-yellow high-purity liquid for research applications.
  • Potent, noncompetitive inhibitor of DNA primase.
  • Blocks primer RNA formation and DNA synthesis.
  • Inhibits ribonucleotide reductase and DNA polymerase.
  • High reported purity suitable for analytical work.
  • Liquid form for rapid dissolution and use.
  • Requires protected, low-temperature storage to maintain stability.
Catalog No. 50-002-21843 Supplier Medchemexpress LLC Supplier No. HY136650A5MG
Fludarabine triphosphate trisodium is the active triphosphate metabolite of fludarabine that inhibits DNA primase and interferes with DNA synthesis. It blocks primer RNA formation, inhibits ribonucleotide reductase and DNA polymerase, and promotes apoptosis in susceptible cells. Supplied as a colorless to light-yellow high-purity liquid for research applications.
  • Potent, noncompetitive inhibitor of DNA primase.
  • Blocks primer RNA formation and DNA synthesis.
  • Inhibits ribonucleotide reductase and DNA polymerase.
  • High reported purity suitable for analytical work.
  • Liquid form for rapid dissolution and use.
  • Requires protected, low-temperature storage to maintain stability.