Learn More
TARGETMOL CHEMICALS INC Fludazonium chloride 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T122641MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.Fludazonium chloride (NSC-351149) is an antifungal agent. Purity 98.09%
Catalog No. 50-287-8350