Learn More
Cayman Chemical Fludioxonil, Purity: ≥98%, CAS: 131341-86-1, 50 mg
SDP
Catalog No. 502934353
Encompass_Preferred
A fungicide; inhibits the growth of B. cinerea isolates from strawberry and blackberry crops (EC50s = 0.06-0.38 mg/L); inhibits the growth of S. cerevisiae AF293 and C. albicans IFO1385; an ERα agonist (EC50 = 3.7 μM in a yeast reporter assay); induces expression of the oncogenic microRNA miR-21, inhibits estradiol-induced proliferation, and reduces cell motility of MCF-7 breast cancer cells in vitro
Catalog No. 50-293-4353 Supplier Cayman Chemical Supplier No. 2581850 MG
A fungicide; inhibits the growth of B. cinerea isolates from strawberry and blackberry crops (EC50s = 0.06-0.38 mg/L); inhibits the growth of S. cerevisiae AF293 and C. albicans IFO1385; an ERα agonist (EC50 = 3.7 μM in a yeast reporter assay); induces expression of the oncogenic microRNA miR-21, inhibits estradiol-induced proliferation, and reduces cell motility of MCF-7 breast cancer cells in vitro