Learn More
Cayman Chemical Fludrocortisone Acetate, Purity: ≥98%, CAS: 514-36-3, 500 mg
SDP
Catalog No. 502933879
Encompass_Preferred
A mineralocorticoid agonist; promotes sodium retention in rats in a dose-dependent manner with 37% of sodium excreted compared with control when used at a dose of 5 µg/animal
Catalog No. 50-293-3879 Supplier Cayman Chemical Supplier No. 24032500 MG
A mineralocorticoid agonist; promotes sodium retention in rats in a dose-dependent manner with 37% of sodium excreted compared with control when used at a dose of 5 µg/animal