Learn More
TARGETMOL CHEMICALS INC FLUDROCORTISONE 10MG
SDP
Catalog No. 502581166
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fludrocortisone (Florinef) is a synthetic corticosteroid with moderate glucocorticoid potency and much greater mineralocorticoid potency. Fludrocortisone can be used to studies on treating low blood pressure. purity: 98%
Catalog No. 50-258-1166 Supplier TARGETMOL CHEMICALS INC Supplier No. T1666L10MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fludrocortisone (Florinef) is a synthetic corticosteroid with moderate glucocorticoid potency and much greater mineralocorticoid potency. Fludrocortisone can be used to studies on treating low blood pressure. purity: 98%