Learn More
Medchemexpress LLC Fludrocortisone acetate | 514-36-3 | 99.6% | 1 ML
SDP
Catalog No. 5000411120
Encompass_Preferred
Fludrocortisone acetate is an orally active synthetic mineralocorticoid. It effectively controls sodium retention and is used in studies of cardiac injury, adrenal insufficiency, and orthostatic hypotension. Research indicates its role in managing conditions related to sodium balance and cardiovascular health.
  • Effectively controls sodium retention
  • Supports research into cardiac injury
  • Supports research into adrenal insufficiency
  • Supports research into orthostatic hypotension
  • Demonstrates efficacy in reducing specific enzyme activities related to cardiac injury
  • Shown to decrease UACR in healthy cats
  • Contributes to restoring quiescent morphology in choroidal endothelial cells
  • Assists in reducing membrane permeability in stimulated choroidal endothelial cells
Catalog No. 50-004-11120 Supplier Medchemexpress LLC Supplier No. HYB1203A10MM/1ML
Fludrocortisone acetate is an orally active synthetic mineralocorticoid. It effectively controls sodium retention and is used in studies of cardiac injury, adrenal insufficiency, and orthostatic hypotension. Research indicates its role in managing conditions related to sodium balance and cardiovascular health.
  • Effectively controls sodium retention
  • Supports research into cardiac injury
  • Supports research into adrenal insufficiency
  • Supports research into orthostatic hypotension
  • Demonstrates efficacy in reducing specific enzyme activities related to cardiac injury
  • Shown to decrease UACR in healthy cats
  • Contributes to restoring quiescent morphology in choroidal endothelial cells
  • Assists in reducing membrane permeability in stimulated choroidal endothelial cells