Learn More
Cayman Chemical Fluensulfon 10mg
SDP
Catalog No. 502986148
Encompass_Preferred
A nematicide; inhibits mobility of P. penetrans, P. thornei, and X. index migratory nematodes at 4 mg/L; reduces the number of P. penetrans and P. thornei nematodes in soil when applied at 2 mg/L before planting crops; metabolized by murine-specific Cyp2f2 in mouse lung club cells and induces species-specific bronchiolar hyperplasias in mice
Catalog No. 50-298-6148 Supplier Cayman Chemical Supplier No. 2155410 MG
A nematicide; inhibits mobility of P. penetrans, P. thornei, and X. index migratory nematodes at 4 mg/L; reduces the number of P. penetrans and P. thornei nematodes in soil when applied at 2 mg/L before planting crops; metabolized by murine-specific Cyp2f2 in mouse lung club cells and induces species-specific bronchiolar hyperplasias in mice