Learn More
Fluotics Fluent Evo LiHa P200 conductive sterile
Catalog No. NC2195951
Encompass
Tecan Fluent Evo LiHa P200, conductive, sterile, Hanging Rack, for FCA/LiHa, working volume: 200 uL, Case Size: 24 racks/case with 96 tips per rack, Compare to: 30000627
This item is not returnable. View return policy
Catalog No. NC2195951 Supplier Fluotics Supplier No. LHA200CB.ST
Tecan Fluent Evo LiHa P200, conductive, sterile, Hanging Rack, for FCA/LiHa, working volume: 200 uL, Case Size: 24 racks/case with 96 tips per rack, Compare to: 30000627