Learn More
Fluotics Fluent Evo LiHa P50 conductive filter/sterile
Catalog No. NC2195978
Encompass
Tecan Fluent Evo LiHa P50, conductive, filter/sterile, Single Stack, refill tray, for FCA/LiHa, working volume: 50 uL, Case Size: 24 racks/case with 96 tips per rack, Compare to: 30057813
This item is not returnable. View return policy
Catalog No. NC2195978 Supplier Fluotics Supplier No. LHA50CBR.F
Tecan Fluent Evo LiHa P50, conductive, filter/sterile, Single Stack, refill tray, for FCA/LiHa, working volume: 50 uL, Case Size: 24 racks/case with 96 tips per rack, Compare to: 30057813