Learn More
Fluotics Fluent Evo LiHa P50 conductive sterile
Catalog No. NC2195981
Encompass
Tecan Fluent Evo LiHa P50, conductive, sterile, Single Stack, refill tray, for FCA/LiHa, working volume: 50 uL, Case Size: 24 racks/case with 96 tips per rack, Compare to: 30057811
This item is not returnable. View return policy
Catalog No. NC2195981 Supplier Fluotics Supplier No. LHA50CBR.ST
Tecan Fluent Evo LiHa P50, conductive, sterile, Single Stack, refill tray, for FCA/LiHa, working volume: 50 uL, Case Size: 24 racks/case with 96 tips per rack, Compare to: 30057811