Learn More
Fluotics Fluent Evo MCA comp P50 filter/sterile
Supplier:  Fluotics MCA50.F
Encompass
Tecan Fluent Evo MCA compatible P50, filter/sterile, for MCA head, working volume: 50 uL, Case Size: 50 racks/case with 96 tips per rack, Compare to: 30038608
Catalog No. NC2195986