Learn More
Fluotics Fluent Evo MCA comp P500 filter/sterile
Catalog No. NC2195993
Encompass
Tecan Fluent Evo MCA compatible P500, filter/sterile, for MCA head, working volume: 430 uL, Case Size: 50 racks/case with 96 tips per rack, Compare to: 30046342
This item is not returnable. View return policy
Catalog No. NC2195993 Supplier Fluotics Supplier No. MCA500.F
Tecan Fluent Evo MCA compatible P500, filter/sterile, for MCA head, working volume: 430 uL, Case Size: 50 racks/case with 96 tips per rack, Compare to: 30046342