Learn More
Fluotics Fluent Evo MCA comp P500 non-sterile
Catalog No. NC2195995
Encompass
Tecan Fluent Evo MCA compatible P500, non-sterile, for MCA head, working volume: 500 uL, Case Size: 50 racks/case with 96 tips per rack, Compare to: 30046341
This item is not returnable. View return policy
Catalog No. NC2195995 Supplier Fluotics Supplier No. MCA500.NS
Tecan Fluent Evo MCA compatible P500, non-sterile, for MCA head, working volume: 500 uL, Case Size: 50 racks/case with 96 tips per rack, Compare to: 30046341