Learn More
Cayman Chemical FlufenamIc AcId-d4 500ug
SDP
Catalog No. 502949879
Encompass_Preferred
An internal standard for the quantification of flufenamic acid by GC- or LC-MS
Catalog No. 50-294-9879 Supplier Cayman Chemical Supplier No. 33272500 UG
An internal standard for the quantification of flufenamic acid by GC- or LC-MS